Would you like to log in again?
Masterflex® High-Viscosity Sanitary Drum Pumps, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
800DD1851P39/A8FP 800DD1851P39/522 800SR1851P39/ENCV
MFLX75220-20EA 12060 USD InStock
MFLX75220-20 MFLX75220-30 MFLX75220-00
Masterflex® High-Viscosity Sanitary Drum Pumps, Avantor®
Pumps Drum/transfer Pumps
Capable of pumping fluids with viscosities up to 10,000 cps.
  • Progressive cavity design allows for low-shear properties
  • Flow rates up to 12 GPM (45 LPM)
  • 316 stainless steel construction with PTFE elastomers
These drum pumps are engineered specifically for transferring food products, pharmaceuticals, and cosmetic products from barrels, kettles, and tanks. They offer a progressive cavity, positive-displacement delivery system resulting in a continuous flow of material with little product degradation. This unique design allows you to quickly dismantle, clean, and re-assemble the pump.
Delivery information: Includes drive and pump tube.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty