Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Masterflex® Submersible Dewatering Centrifugal Pumps, Avantor®

Supplier: Avantor Fluid Handling

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Masterflex®
- EPD-3AS1 OPTIMA-3AS1 EPD-3MS1 7550-74 EPD-7MS1 EPD-7MT4 EPD-5AS1 OPTIMA-3MS1 OPTIMA-3AS1-SLIMFLT
MFLX75504-50EA 539 USD InStock
MFLX75504-50 MFLX75504-60 MFLX75507-52 MFLX75507-74 MFLX75504-55 MFLX75507-76 MFLX75507-70 MFLX75507-50 MFLX75507-72 MFLX75507-60
Masterflex® Submersible Dewatering Centrifugal Pumps, Avantor®
Pumps Centrifugal Pumps
Semi-open impeller design pumps solids up to 3/8" in slurries and up to 81 GPM.

  • Stainless steel construction resists corrosion and rust
  • Pumps with automatic operation have float switch; manual operation models activate with a switch
  • Air-filled motors have permanent split capacitor design with built-in thermal protection and automatic reset

Operate pumps either fully or partially submerged. Pumps are ideal for slurries and dirty or semi-dirty waters.

Delivery information: Includes 20 ft (6.1 m) water-resistant #16 AWG cord.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty