Would you like to log in again?
Abaque™ Peristaltic Natural Rubber Replacement Hose
Supplier: Abaque
Y218938 Y218724 Y218941 Y218727 Y218712 Y218748 Y218736
MFLX75810-26EA 222 USD InStock
MFLX75810-26 MFLX75810-48 MFLX75810-43 MFLX75810-32 MFLX75810-21 MFLX75810-64 MFLX75810-83
Abaque™ Peristaltic Natural Rubber Replacement Hose
Tubing Peristaltic Pump Tubing
Highly resilient with excellent abrasion resistance and strength.
  • All listed hoses in this ad are made from Natural Rubber.
  • The hose IDs are: 10, 15, 20, 25, 32, 40, 50, 65, 80, 100 mm. Each liquid end is designed to accept one particular hose ID version.
The hose of a Peristaltic Hose pump is the only component that comes in contact with the pumped medium. Abaque engineers / manufactures their hoses from high quality compounded rubber, reinforced by layers of nylon. Once constructed, the hose is then machined with precision for efficiency, accuracy, longevity, and optimal compression.
Ideal for applications where the following properties are needed:
Highly resilient, excellent abrasion resistance, the most common hose in peristaltic markets. Generally resistant with lightly corrosive chemical applications, abrasive slurries, etc.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty