Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Stainless Steel Motor Sanitary Centrifugal Pumps

Supplier: BPH PUMP & EQUIPMENT INC

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

AC-216-4.25-X58S AC-114-2.5-J20S AC-114-4.0-Y58S AC-114-4.0-J52S
MFLX76720-00EA 3337 USD InStock
MFLX76720-00 MFLX76720-20 MFLX76720-40 MFLX76720-05
Stainless Steel Motor Sanitary Centrifugal Pumps
Pumps Centrifugal Pumps
Clean-in-place (CIP) 3A sanitary transfer pump.

  • 316L stainless steel liquid end construction
  • Washdown motors available in stainless steel
  • Stainless steel adapters are standard
  • Ideal for RO water and pharmaceutical manufacturing

Piping changes are eliminated due to the interchangeable design that these economical pumps offer when compared to other pumps. Models are available with inlets up to 6" contact our Application Specialists for assistance in sizing the proper pump for your system.

Delivery information: Includes conduit box for hardwiring.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty