Would you like to log in again?
Masterflex® Ismatec® Microflex Variable-Speed Pump Systems, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
77122-34 77122-34 77122-30 77122-30 77122-32 77122-34 77122-34 77122-30 77122-32 77122-34 77122-32 77122-30 77122-34 77122-32 77122-34 77122-32 77122-32 77122-30 77122-30 77122-32 77122-34 77122-30 77122-34 77122-32 77122-32 77122-30 77122-30
MFLX77122-34-UKCS 0 USD InStock
MFLX77122-34-UK MFLX77122-32-UK MFLX77122-30-DK MFLX77122-34-IL MFLX77122-32-IL MFLX77122-34-CH MFLX77122-34-IN MFLX77122-32-IN MFLX77122-32-CH MFLX77122-34-IT MFLX77122-32-IT MFLX77122-32 MFLX77122-34-EU MFLX77122-34 MFLX77122-30-AU MFLX77122-32-EU MFLX77122-34-AU MFLX77122-30-EU MFLX77122-32-AU MFLX77122-30-IT MFLX77122-30 MFLX77122-30-IN MFLX77122-30-CH MFLX77122-30-IL MFLX77122-34-DK MFLX77122-32-DK MFLX77122-30-UK
Masterflex® Ismatec® Microflex Variable-Speed Pump Systems, Avantor®
Pumps Peristaltic Pumps
Easy-to-use low-flow pumps feature reversible pumping, remote start/stop, and rapid tubing changes.
  • Polyphenylene sulfide (PPS), acetal, and copolyester pump head with four stainless steel (SS) rollers
  • Pumps accept continuous lengths of tubing for a clean, uninterrupted flow path with no fittings.
  • Separate single-turn speed control and cw/off/ccw switch with green LED power indicator—maintain speed setting when you turn pump on/off or reverse direction.
  • Reversible permanent magnet DC motor; purge fluid before or after pumping; pump fluid in either direction.
  • Remote control connections on back of pump; start/stop pump with a contact closure.
These easy-to-use pumps feature reversible pumping, remote control capabilities, and rapid tubing changes. Ideal for consistent fluid delivery in analyzer and other low-flow laboratory pumping applications. The quick-loading Microflex pump head has average fixed occlusion for tubing; no occlusion adjustments required.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty