Would you like to log in again?
Masterflex® L/S® Compact Variable-Speed Drive, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
77200-20 77200-20 77200-20 77200-20 77200-12 77200-20 77200-20 77200-20 77200-20 77200-20
MFLX77200-20-DKCS 0 USD InStock
MFLX77200-20-DK MFLX77200-20 MFLX77200-20-CH MFLX77200-20-IN MFLX77200-20-UK MFLX77200-20-IL MFLX77200-20-AU MFLX77200-20-EU MFLX77200-20-IT MFLX77200-12
Masterflex® L/S® Compact Variable-Speed Drive, Avantor®
Pumps Pump Drives Peristaltic Pump Drives
Stackable, compact pump drive delivers versatile performance in limited space.
  • Flow rate 2.1 to 560 ml/min—flow rate depends on drive rpm, pump head, and tubing size
  • Stackable, compact drive is perfect for small work areas
  • Constructed with a painted steel housing with IP22 rating
  • No difficult programming—single switch for forward/off/reverse and analog controller
  • Pump in either direction—also allows purging before or after pumping
  • Remote control of start/stop via contact closure
These compact drives are a perfect when space is limited, as they fit just about anywhere. The variable-speed design makes them ideal for when you need to pump fluid at a higher rate. Drives accept one Easy-Load® or standard pump head. Use drives with L/S® Precision tubing—accepts sizes L/S 13, L/S 14, L/S 16, L/S 25, and L/S 17. Only silicone and C-Flex® tubing formulations are recommended for use with L/S 25 and L/S 17 sizes.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty