Would you like to log in again?
Masterflex® L/S® Compact Variable-Speed Drives, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
77240-30 77240-30 77240-30 77240-20 77240-20 77240-20 77240-30 77240-30 77240-20 77240-20 77240-20 77240-30 77240-30 77240-30 77240-20 77240-20 77240-30 77240-20
MFLX77240-30-EUCS 0 USD InStock
MFLX77240-30-EU MFLX77240-20-IL MFLX77240-30-IN MFLX77240-30-CH MFLX77240-30 MFLX77240-20-UK MFLX77240-30-DK MFLX77240-20 MFLX77240-30-IT MFLX77240-20-EU MFLX77240-20-AU MFLX77240-30-IL MFLX77240-30-AU MFLX77240-20-DK MFLX77240-20-IN MFLX77240-30-UK MFLX77240-20-CH MFLX77240-20-IT
Masterflex® L/S® Compact Variable-Speed Drives, Avantor®
Pumps Peristaltic Pumps
Make low-flow pumping easy!
  • Flow rate 1.8 to 220 ml/min with L/S tubing—flow rate depends on drive rpm, pump head, and tubing size
  • Pump head accepts continuous lengths of tubing for clean flow paths
  • Dual-channel model delivers synchronous flow from two separate channels; single-channel model delivers highest flow rate
  • Automatic tubing retention makes tube loading quick and easy
  • Separate single-turn speed control and forward/off/reverse switch—maintain speed setting when turning pump on or off
  • Remote start/stop via contact closure on back of pump
These easy-to-use complete pumps offer a wide occlusion angle, which prevents fluid backflow up to the rated pressure of the tubing. Prime or flush tubing quickly by depressing the MAX button. Self-lubricating thermoplastic polyester rollers are low friction and long wearing for smooth operation and long life.
This compact, stackable pump features a durable steel housing with chip-resistant epoxy-based paint. Wide occlusion angle prevents fluid backflow up to the rated pressure of the tubing. MAX button runs motor at full speed to rapidly prime or flush tubing. Self-lubricating thermoplastic polyester (Ertalyte®TX) rollers are low friction and long wearing for smooth operation and long life.
Note:Dual-channel model must have same size tubing loaded in both channels to ensure tubing retention.
Delivery information: Includes L/S® Miniflex® pump head and dual-voltage (115/230 VAC) benchtop power supply with IEC320/CEE22 socket. Pump is shipped with cord/plug assembly specific to country of destination. Tubing is not included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty