Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Masterflex® L/S® Digital Modular Dispensing Drives, Avantor®

Supplier: Avantor Fluid Handling

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Masterflex®
77301-60 77301-60 77301-40 77301-70 77301-70 77301-40 77301-70 77301-70 77301-60 77301-70 77301-50 77301-40 77301-40 77301-60 77301-60 77301-40 77301-70 77301-50 77301-70 77301-50 77301-50 77301-50 77301-60 77301-40 77301-40 77301-60 77301-60 77301-50 77301-40 77301-40 77301-50 77301-50 77301-70 77301-50 77301-70 77301-60
MFLX77301-50-DKCS 0 USD InStock
MFLX77301-50-DK MFLX77301-70-IL MFLX77301-60-UK MFLX77301-70-CH MFLX77301-70-IN MFLX77301-40-AU MFLX77301-40-EU MFLX77301-60-IL MFLX77301-60-IN MFLX77301-60-CH MFLX77301-50-AU MFLX77301-70-UK MFLX77301-40-DK MFLX77301-60 MFLX77301-40 MFLX77301-50-EU MFLX77301-60-AU MFLX77301-50-CH MFLX77301-50-IN MFLX77301-50-IT MFLX77301-40-UK MFLX77301-70-DK MFLX77301-60-IT MFLX77301-50-IL MFLX77301-60-EU MFLX77301-50-UK MFLX77301-70 MFLX77301-70-AU MFLX77301-40-IT MFLX77301-40-IN MFLX77301-40-CH MFLX77301-60-DK MFLX77301-70-IT MFLX77301-40-IL MFLX77301-50 MFLX77301-70-EU
Masterflex® L/S® Digital Modular Dispensing Drives, Avantor®
Pumps Pump Drives Peristaltic Pump Drives
Precision digital drives offer three dispensing modes and precision low-maintenance motors— modular format lets you place motor and controller where convenient.

  • Flow rate 0.0012 to 3400 ml/min with L/S —flow rate depends on drive rpm, pump head, and tubing size
  • Digital interface with graphical LCD shows pump performance and operating mode— continuous run, timed dispense, copy dispense, or volume dispense
  • Remote control available using analog communication—voltage, current and contact closure
  • Select your preferred language from the user-friendly menu for programming and operation
  • Dispense by Volume in ml (0.001 to 9,999 mL), L, or gal; Copy—1 to 9,999 dispense cycles; or Time—time interval of on/off cycles (1 sec to 99 hr, 59 min, 59 sec per cycle)
  • Antidrip function ensures dispensing accuracy
  • Brushless, maintenance-free motor offers ±0.1% speed control accuracy with a 6000 to 1 turndown ratio
  • Reversible motor—purge tubing before or after pumping or pump fluid in either direction
  • Powerful motors let you stack pump heads for increased flow rates and for multichannel applications

These precision drives feature a programmable dispense interval that lets you set the delay between dispense cycles for convenient, automated dispensing by time or by volume. Programmed calibration ensures dispense and display accuracy—drive stores one calibration value per tubing size. Program the drive via the simple membrane keypad; keypad lock/unlock feature protects settings.

Delivery information: Includes 6-ft (1.8-m) line cord with IEC 320/CEE22 socket. These models are shipped with country-specific plug/cord set; please specify ultimate destination when ordering.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty