Would you like to log in again?
Masterflex® L/S Digital Dispensing Systems with Easy Load II Pump Heads, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
77714-77 77714-78 77714-80 77714-80 77714-79 77714-76 77714-76 77714-79 77714-77 77714-80 77714-77 77714-78 77714-81 77714-79 77714-79 77714-81 77714-79 77714-78 77714-77 77714-79 77714-78 77714-76 77714-77 77714-77 77714-81 77714-81 77714-78 77714-76 77714-80 77714-77 77714-80 77714-79 77714-77 77714-78 77714-76 77714-81 77714-81 77714-80 77714-79 77714-80 77714-76 77714-78 77714-76 77714-76 77714-81 77714-80 77714-80 77714-76 77714-81 77714-78 77714-77 77714-78 77714-81 77714-79
MFLX77714-76-ITCS 0 USD InStock
MFLX77714-76-IT MFLX77714-76-EU MFLX77714-79-UK MFLX77714-80-CH MFLX77714-76-AU MFLX77714-81-DK MFLX77714-81-IL MFLX77714-79-IL MFLX77714-80 MFLX77714-79-IN MFLX77714-81 MFLX77714-77-CH MFLX77714-78-DK MFLX77714-81-UK MFLX77714-79-IT MFLX77714-81-EU MFLX77714-79-EU MFLX77714-79-AU MFLX77714-78-UK MFLX77714-81-IT MFLX77714-81-IN MFLX77714-80-DK MFLX77714-78-IL MFLX77714-78-IN MFLX77714-80-UK MFLX77714-76-CH MFLX77714-81-AU MFLX77714-77-DK MFLX77714-78-IT MFLX77714-80-IT MFLX77714-78-EU MFLX77714-80-EU MFLX77714-80-IN MFLX77714-78-AU MFLX77714-77-UK MFLX77714-80-IL MFLX77714-77-IL MFLX77714-79-CH MFLX77714-77-IN MFLX77714-80-AU MFLX77714-76-DK MFLX77714-77-IT MFLX77714-77-EU MFLX77714-77-AU MFLX77714-76-UK MFLX77714-81-CH MFLX77714-77 MFLX77714-78 MFLX77714-79 MFLX77714-78-CH MFLX77714-76-IL MFLX77714-79-DK MFLX77714-76-IN MFLX77714-76
Masterflex® L/S Digital Dispensing Systems with Easy Load II Pump Heads, Avantor®
Pumps Peristaltic Pumps
Precision digital drive offers a maintenance-free high-accuracy motor and a graphical interface – makes setup and operation easier than ever.
  • Available in an IP33 plastic enclosure or IP65 stainless steel enclosure
  • Antidrip function ensures dispensing accuracy
  • Open head sensor
Graphical LCD shows four operating modes continuous run, timed dispense, copy dispense, volume dispense
These precision drives offer analog remote control options and simple programming—ideal for automated process applications. Drives feature a programmable dispense interval that lets you set the delay between dispense cycles for convenient, automated dispensing. Programmed calibration ensures dispense and display accuracy—drive stores one calibration value per tubing size.
Program the drive via the simple membrane keypad; keypad lock/unlock feature protects settings. Motor is reversible so you can purge tubing before or after pumping and pump fluid in either direction.
View pump performance continually.
Certifications: ETL, cETL, CE marked.
Ordering information: Includes NEMA 3-prong ground power cable. Accessories not included.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty