Would you like to log in again?
Masterflex® L/S® Analog Variable-Speed Console Drive Systems, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
77910-45 77910-65 77910-70 77910-55 77910-50 77910-45 77910-65 77910-65 77910-55 77910-75 77910-75 77910-55 77910-65 77910-55 77910-65 77910-75 77910-75 77910-45 77910-75 77910-75 77910-60 77715-91 77910-45 77910-65 77910-55 77910-45 77910-65 77910-65 77910-75 77910-55 77910-55 77910-45 77910-55 77910-75 77910-45 77910-45 77910-65 77910-45 77715-90 77910-75 77910-55 77910-40
MFLX77910-55-ILCS 0 USD InStock
MFLX77910-55-IL MFLX77910-65-EU MFLX77910-75-AU MFLX77715-90 MFLX77715-91 MFLX77910-65-AU MFLX77910-75-EU MFLX77910-55-CH MFLX77910-55-IN MFLX77910-45-UK MFLX77910-55-IT MFLX77910-75-IT MFLX77910-45-IL MFLX77910-45-IN MFLX77910-45-CH MFLX77910-75-DK MFLX77910-55 MFLX77910-75 MFLX77910-45-IT MFLX77910-50 MFLX77910-55-UK MFLX77910-65-DK MFLX77910-70 MFLX77910-45-AU MFLX77910-75-UK MFLX77910-45-EU MFLX77910-55-DK MFLX77910-65-UK MFLX77910-55-EU MFLX77910-55-AU MFLX77910-45 MFLX77910-75-IN MFLX77910-75-CH MFLX77910-65-IL MFLX77910-65 MFLX77910-75-IL MFLX77910-65-IN MFLX77910-65-CH MFLX77910-40 MFLX77910-60 MFLX77910-45-DK MFLX77910-65-IT
Masterflex® L/S® Analog Variable-Speed Console Drive Systems, Avantor®
Pumps Peristaltic Pumps
Compact, cost effective pump systems for general fluid transfer applications.
  • Simple operation with single-turn speed control
  • Separate power switch enables you to maintain speed setting when turning drive on/off; front-panel LED indicates power on
  • Reversible motor lets you easily prime/purge tubing and pump in either direction
  • Remote control of start/stop via DB9 connector on back of drive
Choose a convenient, complete system all with one catalog number.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty