Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 25, 2024. During this time, our site will be not available. We appreciate your patience.
Masterflex® L/S® PTFE-Diaphragm Pump System, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
77915-10 77915-10 77915-10 77915-10 77915-10 77915-10 77915-10 77915-10 77915-10
MFLX77915-10-EUCS 0 USD InStock
MFLX77915-10-EU MFLX77915-10-IL MFLX77915-10-AU MFLX77915-10-CH MFLX77915-10-IN MFLX77915-10 MFLX77915-10-UK MFLX77915-10-DK MFLX77915-10-IT
Masterflex® L/S® PTFE-Diaphragm Pump System, Avantor®
Pumps Diaphragm Pumps
Ideal for chemical feed/metering and applications requiring high-purity, high-pressure or both.
  • Flow range 80 to 800 ml/min—flow rate depends on drive rpm and tubing size
  • PTFE wetted parts help maintain fluid purity
  • Pump through filters or into pressurized system
  • Check valve on inlet side of pump holds prime for up to two hours when pump is off
  • Unique geometry minimizes fluid churning and dead volume for high metering accuracy without reagent contamination
  • Accurate and repeatable settings with 3-digit LED display of rpm
  • Maximize processes in a minimal footprint by stacking splash-resistant, IP33-rated drives
  • Two ways to control speed, direction, and start/stop—use either the membrane keypad or remote-control capabilities
These L/S® precision variable-speed console drives offer precise ±0.25% speed control for accurate flow regulation. The large LED display and LED indicators confirm motor rpm and motor direction at a glance. The complete pump systems include all you need to start pumping.
Delivery information: Includes pTFE-diaphragm pump head 07090-42, 12 ft (3.6-m) of PTFE tubing (1/4" ID x 3/8" OD), L/S drive 07528-10, two PFA pipe adapters (1/4" NPT(F) x 3/8" OD), foot valve, mounting tools, and coupling hardware.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty