Would you like to log in again?
Masterflex® L/S® Precision Process Pump Systems, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
77916-50 77916-52 77916-42 77916-47 77916-42 77916-47 77916-47 77916-40 77916-47 77916-42 77916-47 77916-50 77916-52 77916-40 77916-42 77916-50 77916-42 77916-40 77916-47 77916-57 77916-52 77916-55 77916-42 77916-40 77916-50 77916-47 77916-52 77916-50 77916-40 77916-42 77916-52 77916-40 77916-42 77916-52 77916-52 77916-45 77916-47 77916-50 77916-47 77916-52 77916-40 77916-50 77916-42 77916-40 77916-52 77916-50 77916-50 77916-40
MFLX77916-47-AUCS 0 USD InStock
MFLX77916-47-AU MFLX77916-50-UK MFLX77916-52 MFLX77916-57 MFLX77916-52-UK MFLX77916-55 MFLX77916-52-IL MFLX77916-47-IT MFLX77916-42-DK MFLX77916-40-DK MFLX77916-47-EU MFLX77916-50-DK MFLX77916-52-DK MFLX77916-47-DK MFLX77916-42-IL MFLX77916-42-UK MFLX77916-40-UK MFLX77916-50 MFLX77916-42 MFLX77916-40-IT MFLX77916-42-EU MFLX77916-40 MFLX77916-50-AU MFLX77916-47 MFLX77916-40-EU MFLX77916-42-AU MFLX77916-47-UK MFLX77916-52-AU MFLX77916-45 MFLX77916-40-IL MFLX77916-42-IN MFLX77916-42-CH MFLX77916-40-CH MFLX77916-40-IN MFLX77916-42-IT MFLX77916-47-IN MFLX77916-47-CH MFLX77916-52-IT MFLX77916-47-IL MFLX77916-50-CH MFLX77916-50-IN MFLX77916-52-CH MFLX77916-52-IN MFLX77916-50-IL MFLX77916-50-EU MFLX77916-40-AU MFLX77916-52-EU MFLX77916-50-IT
Masterflex® L/S® Precision Process Pump Systems, Avantor®
Pumps Peristaltic Pumps
Easy-to-use drives offer precise speed control and digital display of rpm—NEMA 4X housings stand up to process and washdown applications.
  • Four digit LED display shows motor rpm—confirm speed setting at a glance
  • Integrated open-head sensor stops drive motor when pump head is opened
  • Reversible motor—purge tubing before or after pumping; pump fluid in either direction
  • LED indicators confirm motor direction
  • Powerful motors let you stack 2 or 4 pump heads for increased flow rates and for multichannel applications
  • Separate speed and power (on/off) control—maintain speed setting while turning pump on/off
  • NEMA 4X (IP66) rated housing
  • Remote control capabilities via a watertight connector on back of drive
These accurate console drives have powerful motors, speed display, and ±0.25% speed control for precise flow regulation—ideal for metering applications. Remote speed control requires cable assembly 77300-32.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty