Would you like to log in again?
Masterflex® L/S® Digital Modular Dispensing Pump Systems, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
77923-80 77923-90 77923-80 77923-80 77923-90 77923-90 77923-80 77923-90 77923-80 77923-90 77923-90 77923-80 77923-90 77923-80 77923-90 77923-80 77923-80 77923-90
MFLX77923-80-EUCS 0 USD InStock
MFLX77923-80-EU MFLX77923-80-IT MFLX77923-90-UK MFLX77923-80-DK MFLX77923-80-CH MFLX77923-80-IN MFLX77923-80 MFLX77923-80-IL MFLX77923-90-AU MFLX77923-90 MFLX77923-90-EU MFLX77923-90-IT MFLX77923-90-DK MFLX77923-80-UK MFLX77923-90-IL MFLX77923-90-CH MFLX77923-90-IN MFLX77923-80-AU
Masterflex® L/S® Digital Modular Dispensing Pump Systems, Avantor®
Pumps Peristaltic Pumps
Precision digital drives offer three dispensing modes and precision low-maintenance motors— modular format lets you place motor and controller where convenient.
  • Digital interface with graphical LCD shows pump performance and operating mode— continuous run, timed dispense, copy dispense, or volume dispense
  • Remote control available using analog communication—voltage, current and contact closure
  • Select your preferred language from the user-friendly menu for programming and operation
  • Dispense by Volume in ml (0.001 to 9,999 mL), L, or gal; Copy—1 to 9,999 dispense cycles; or Time—time interval of on/off cycles (1 sec to 99 hr, 59 min, 59 sec per cycle)
  • Antidrip function ensures dispensing accuracy
  • Brushless, maintenance-free motor offers ±0.1% speed control accuracy with a 6000 to 1 turndown ratio
  • Reversible motor—purge tubing before or after pumping or pump fluid in either direction
  • Powerful motors let you stack pump heads for increased flow rates and for multichannel applications
These precision drives feature a programmable dispense interval that lets you set the delay between dispense cycles for convenient, automated dispensing by time or by volume. Precise PWM speed control (±0.1%) maximizes accuracy and efficiency while the brushless motor gives you quiet, maintenance-free reliability. Programmed calibration ensures dispense and display accuracy—drive stores one calibration value per tubing size. Program the drive via the simple membrane keypad; keypad lock/unlock feature protects settings.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty