Would you like to log in again?
Masterflex® L/S® Variable Speed Analog Console Pump Systems, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
77925-10 77925-10 77925-10 77926-10 77926-10 77926-10 77926-10 77916-10 77916-20 77916-20 77925-10 77916-10 77916-10 77925-10 77926-10 77916-10 77916-10 77916-20 77926-10 77926-10 77925-10 77916-20 77925-10 77916-10 77916-10 77916-20 77916-20 77925-10 77926-10 77916-20 77916-10 77916-20 77925-10 77916-10 77926-10 77927-10 77916-20
MFLX77926-10-INCS 0 USD InStock
MFLX77926-10-IN MFLX77916-10-CH MFLX77926-10-CH MFLX77916-10-IN MFLX77916-10 MFLX77926-10-IL MFLX77916-20-AU MFLX77916-10-IL MFLX77925-10-DK MFLX77916-20-EU MFLX77916-20-IT MFLX77916-20-IN MFLX77916-20-CH MFLX77916-10-UK MFLX77926-10-UK MFLX77925-10-EU MFLX77927-10 MFLX77925-10-AU MFLX77925-10 MFLX77925-10-IT MFLX77916-10-DK MFLX77926-10-DK MFLX77916-20 MFLX77925-10-IL MFLX77925-10-IN MFLX77925-10-CH MFLX77916-20-DK MFLX77916-10-AU MFLX77926-10-AU MFLX77916-20-IL MFLX77916-10-EU MFLX77926-10 MFLX77926-10-EU MFLX77925-10-UK MFLX77916-10-IT MFLX77926-10-IT MFLX77916-20-UK
Masterflex® L/S® Variable Speed Analog Console Pump Systems, Avantor®
Pumps Peristaltic Pumps
Don’t lose control of your metering applications—digital display gives you precise control.
  • Accurate and repeatable settings with 3-digit LED display of rpm
  • Pump fluid in either direction with reversible motor—perfect for purging tubing
  • Choose multichannel systems for synchronous flow from all channels
  • Maximize processes in a minimal footprint by stacking splash-resistant, IP33-rated drives
  • Two ways to control speed, direction, and start/stop—use either the membrane keypad or remote-control capabilities
These L/S® precision variable-speed console drives offer precise ±0.25% speed control for accurate flow regulation. The large LED display and LED indicators confirm motor rpm and motor direction at a glance. The complete pump systems include all you need to start pumping.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty