Would you like to log in again?
Masterflex® I/P® Digital Precision Modular Drive Systems, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
77970-50 77970-50 77970-50 77970-40 77970-40 77970-50 77970-40 77970-40 77970-50 77970-40 77970-50 77970-40 77970-40 77970-40 77970-50 77970-40 77970-50 77970-50
MFLX77970-50-DKCS 0 USD InStock
MFLX77970-50-DK MFLX77970-50-IT MFLX77970-40-EU MFLX77970-50-IL MFLX77970-40-UK MFLX77970-50-CH MFLX77970-40-AU MFLX77970-50-IN MFLX77970-50-UK MFLX77970-40-DK MFLX77970-40-IT MFLX77970-40 MFLX77970-50-EU MFLX77970-40-CH MFLX77970-40-IN MFLX77970-50-AU MFLX77970-40-IL MFLX77970-50
Masterflex® I/P® Digital Precision Modular Drive Systems, Avantor®
Pumps Peristaltic Pumps
Place motor and controller where convenient with modular design.
  • Flow rate 0.0006 to 26 LPM with I/P tubing—flow rate depends on drive rpm, pump head, number of pump heads, and tubing size
  • Digital interface with graphical LCD shows pump performance and operating mode— continuous run, timed dispense, copy dispense, or volume dispense
  • Remote control available using analog communication—voltage, current and contact closure
  • Select your preferred language from the user-friendly menu for programming and operation
  • Dispense by Volume in ml (0.001 to 9,999 mL), L, or gal; Copy—1 to 9,999 dispense cycles; or Time—time interval of on/off cycles (1 sec to 99 hr, 59 min, 59 sec per cycle)
  • Antidrip function ensures dispensing accuracy
  • Brushless, maintenance-free motor offers ±0.1% speed control accuracy with a 6000 to 1 turndown ratio
  • Reversible motor—purge tubing before or after pumping and pump fluid in either direction
  • Powerful motor lets you stack two pump heads for increased flow rates and for multichannel applications
These I/P® digital pumps give you rugged performance and precise digital control. These full featured drives are ideal for fluid transfer, dispensing, and metering applications in laboratory, process, and industrial environments including pharmaceutical, food & beverage, dairy, chemical, manufacturing, and water treatment.
Precision drives feature a programmable dispense interval that lets you set the delay between dispense cycles for convenient, automated dispensing by time or by volume. Programmed calibration ensures dispense and display accuracy—drive stores one calibration value per tubing size. Program the drive via the simple membrane keypad; keypad lock/unlock feature protects settings. Select tubing size and enter desired flow rate—drive locks in required rpm.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty