Would you like to log in again?
Masterflex® L/S® Digital Process Pump Systems, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
77975-27 77975-49 77975-25 77975-45 77975-29 77975-45 77975-45 77975-29 77975-29 77975-25 77975-45 77975-27 77975-45 77975-29 77975-45 77975-27 77975-25 77975-27 77975-45 77975-25 77975-29 77975-47 77975-45 77975-29 77975-25 77975-29 77975-25 77975-25 77975-27 77975-27 77975-29 77975-27 77975-45 77975-25 77975-29 77975-27 77975-27 77975-25
MFLX77975-29-DKCS 0 USD InStock
MFLX77975-29-DK MFLX77975-27-DK MFLX77975-29 MFLX77975-49 MFLX77975-27 MFLX77975-25-AU MFLX77975-45-UK MFLX77975-27-AU MFLX77975-45-IT MFLX77975-25-DK MFLX77975-45-IN MFLX77975-45-CH MFLX77975-45-IL MFLX77975-27-IL MFLX77975-29-CH MFLX77975-29-IN MFLX77975-27-IN MFLX77975-27-CH MFLX77975-29-IT MFLX77975-27-IT MFLX77975-29-EU MFLX77975-25-UK MFLX77975-45-EU MFLX77975-27-EU MFLX77975-45-AU MFLX77975-29-AU MFLX77975-25-EU MFLX77975-29-UK MFLX77975-27-UK MFLX77975-45-DK MFLX77975-25-IT MFLX77975-25 MFLX77975-47 MFLX77975-45 MFLX77975-25-CH MFLX77975-25-IN MFLX77975-25-IL MFLX77975-29-IL
Masterflex® L/S® Digital Process Pump Systems, Avantor®
Pumps Peristaltic Pumps
Transfer, dispense, and meter in lab, process and washdown environments.
  • Digital interface with graphical LCD shows pump performance and operating mode— continuous run, timed dispense, copy dispense, or volume dispense
  • Remote control available using analog communication—voltage, current and contact closure
  • Dispense by Volume in ml (0.001 to 9,999 mL), L, or gal; Copy—1 to 9,999 dispense cycles; or Time—time interval of on/off cycles (1 sec to 99 hr, 59 min, 59 sec per cycle)
  • Antidrip function ensures dispensing accuracy
  • Models with open-head sensor stops the drive when the pump head is opened (when used with sensor-equipped pump head)
  • NEMA 4X (IP66) rated housing for washdown applications
  • Models with open-head sensor stop drive motor when pump head is opened
These precision drives offer analog remote control options and simple programming—ideal for automated process applications. Drives feature a programmable dispense interval that lets you set the delay between dispense cycles for convenient, automated dispensing. Programmed calibration ensures dispense and display accuracy—drive stores one calibration value per tubing size. Program the drive via the simple membrane keypad; keypad lock/unlock feature protects settings. Motor is reversible so you can purge tubing before or after pumping and pump fluid in either direction.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty