Would you like to log in again?
Masterflex® Ismatec® Reglo Independent Channel Control (ICC) Peristaltic Pumps, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
ISM4412 ISM4312 ISM4208 ISM4206 ISM4212 ISM4406 ISM4412 ISM4306 ISM4212 ISM4406 ISM4406 ISM4206 ISM4212 ISM4406 ISM4312 ISM4308 ISM4306 ISM4206 ISM4212 ISM4206 ISM4406 ISM4212 ISM4408 ISM4308 ISM4308 ISM4208 ISM4308 ISM4206 ISM4308 ISM4212 ISM4206 ISM4412 ISM4412 ISM4408 ISM4412 ISM4408 ISM4208 ISM4306 ISM4412 ISM4212 ISM4408 ISM4208 ISM4412 ISM4206 ISM4306 ISM4412 ISM4408 ISM4308 ISM4312 ISM4312 ISM4312 ISM4308 ISM4306 ISM4206 ISM4208 ISM4408 ISM4208 ISM4208 ISM4408 ISM4206 ISM4408 ISM4208 ISM4312 ISM4306 ISM4406 ISM4408 ISM4312 ISM4208 ISM4308 ISM4406 ISM4212 ISM4406 ISM4312 ISM4312 ISM4406 ISM4306 ISM4308 ISM4306 ISM4306 ISM4412 ISM4212
MFLX78001-72-DKCS 0 USD InStock
MFLX78001-72-DK MFLX78001-58-EU MFLX78001-60-AU MFLX78001-82-UK MFLX78001-60-EU MFLX78001-58-AU MFLX78001-58-IT MFLX78001-58-IL MFLX78001-62-DK MFLX78001-70-AU MFLX78001-82-IN MFLX78001-58-IN MFLX78001-70-EU MFLX78001-82-IL MFLX78001-80-CH MFLX78001-82-IT MFLX78001-72-IN MFLX78001-80-EU MFLX78001-62-UK MFLX78001-70-CH MFLX78001-80-AU MFLX78001-58-UK MFLX78001-72-IL MFLX78001-68-CH MFLX78001-72-IT MFLX78001-78 MFLX78001-82 MFLX78001-60-CH MFLX78001-62-IN MFLX78001-72-UK MFLX78001-58-DK MFLX78001-62-IL MFLX78001-80 MFLX78001-62-EU MFLX78001-78-CH MFLX78001-82-DK MFLX78001-62-IT MFLX78001-62-AU MFLX78001-78-DK MFLX78001-68 MFLX78001-80-UK MFLX78001-70-DK MFLX78001-72 MFLX78001-72-AU MFLX78001-80-IL MFLX78001-82-CH MFLX78001-72-EU MFLX78001-68-UK MFLX78001-58-CH MFLX78001-70 MFLX78001-68-DK MFLX78001-80-IN MFLX78001-80-IT MFLX78001-60-DK MFLX78001-72-CH MFLX78001-60-UK MFLX78001-70-IL MFLX78001-78-IT MFLX78001-82-EU MFLX78001-70-IN MFLX78001-68-IL MFLX78001-82-AU MFLX78001-68-IN MFLX78001-78-AU MFLX78001-70-IT MFLX78001-58 MFLX78001-78-EU MFLX78001-60-IN MFLX78001-68-EU MFLX78001-60 MFLX78001-62 MFLX78001-60-IL MFLX78001-62-CH MFLX78001-68-AU MFLX78001-78-UK MFLX78001-78-IN MFLX78001-80-DK MFLX78001-78-IL MFLX78001-60-IT MFLX78001-70-UK MFLX78001-68-IT
Masterflex® Ismatec® Reglo Independent Channel Control (ICC) Peristaltic Pumps, Avantor®
Pumps Peristaltic Pumps
Independently control flow and direction of each channel via keypad or your PC.
  • Flow rate 0.001 to 43 ml/min per channel - flow rate depends on drive rpm, rollers, and tubing size
  • USB interface for quick connection to a PC
  • Flexibility of bidirectional flow in each channel
These multi-channel peristaltic pumps let you individually control the flow and direction of each channel from the intuitive keypad or via your computer—expanding your benchtop pumping capabilities with a single compact unit! Pump delivers continuous pumping or precision dispensing with the capability of bidirectional flow in each channel. Plus, independent channel calibration minimizes tube-to-tube differences, resulting in high calibration accuracy.
Order the three-stop tubing size and formulation to suit your application needs (sold separately). More than 100 options exist — the most popular sizes and formulations can be found under Accessories below
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty