Would you like to log in again?
Masterflex® USB A Serial Adapter Cables, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
78006-90 78006-91
MFLX78006-91EA 101 USD InStock
MFLX78006-91 MFLX78006-90
Masterflex® USB A Serial Adapter Cables, Avantor®
Cables
Optimize performance of your Masterflex® MasterSense™ or Ismatec® Reglo Touchscreen pump.
  • Connect your drive via USB A for serial control
Not your standard USB cable. Use one of these cables to connect a Masterflex® MasterSense™ and Ismatec® Reglo Touchscreen pumps to a PC or PLC for serial (RS232) communicaiton. Cables features special capability to support serial interface communication. Chose USB A to USB A for communication to computers. The three wire cable is desinged for serial communcation with PLC applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty