Would you like to log in again?
Pulsafeeder Manual Control Metering Pumps
Supplier: EQUIP SOLUTIONS
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
LPH7SA-WTC3-XXX LPG5SA-VTC3-XXX LD54SA-VTC1-XXX LD04SA-VTC1-XXX LPH8SA-WTCB-XXX LD03SA-VTC1-XXX LD02SA-VTC1-XXX
MFLX78118-00EA 594 USD InStock
MFLX78118-00 MFLX78118-10 MFLX78118-20 MFLX78118-30 MFLX74142-30 MFLX74142-10 MFLX74142-20
Pulsafeeder Manual Control Metering Pumps
Pumps Diaphragm Pumps
Reliable metering performance – guided dual check valves provide precise seating and excellent priming and suction lift.
  • PVC and PTFE construction with external control
  • Ideal for water treatment and chemical feed applications
Manual control pumps feature adjustable stroke rate and length, reliable timing circuits, and circuit protection from voltage and current upsets. Leak-free bleed valves provide safe and easy priming. Pumps are water resistant and can be installed outdoors.
Delivery information: Includes injection/back pressure valve assembly, foot valve/strainer assembly, three-function valve, 4-ft clear PVC tubing, and 8-ft rigid polyethylene discharge tubing. All 115 VAC pumps include a 6-ft power cord with US plug; 230 VAC pumps include a 6-ft power.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty