Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Masterflex® Transfer Tubing, Kynar Flex™ 2750 PVDF, Avantor®

Supplier: Avantor Fluid Handling

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Masterflex®
F-PVDF-625X750-50 F-PVDF-062X125-50 F-PVDF-187X250-50 F-PVDF-500X625-50 F-PVDF-250X375-50 F-PVDF-125X250-50 F-PVDF-312X375-50 F-PVDF-125X187-50 F-PVDF-875X1000-50 F-PVDF-250X312-50 F-PVDF-375X500-50
MFLX95100-03EA 129 USD InStock
MFLX95100-03 MFLX95100-15 MFLX95100-05 MFLX95100-17 MFLX95100-07 MFLX95100-19 MFLX95100-09 MFLX95100-21 MFLX95100-11 MFLX95100-01 MFLX95100-13
Masterflex® Transfer Tubing, Kynar Flex™ 2750 PVDF, Avantor®
Tubing Transfer Tubing
Excellent chemical resistance and UV stable.

  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application

Applications: High purity and temperature, UV sensitive, or aggressive chemical fluid transfer.
Characteristics: Excellent chemical resistance, low TOC, rigid tubing works well with compression and over-molded fittings and adapters. Translucent.
Certification: FDA complaint (21 CFR 177.2600), meets USP Class VI requirements, and is made in accordance to cGMP.
Temperature range: -40 to 275 °F (-40 to 135 °C)
Sterilize: Gamma irradiation, or ethylene oxide.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty