Would you like to log in again?
Masterflex® Transfer Tubing, DEHP-Free Flexible PVC, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
436901705CPI 4362605CP 43690205CP 436901805CP 4361105CP 4360805CP 4361305CP 4361005CP 4360605CP 4360205CP 4360705CP 4363105CP 4360305CP 4360905CP 4361805CP
MFLX95505-02EA 43.9 USD InStock
MFLX95505-02 MFLX95505-24 MFLX95505-14 MFLX95505-00 MFLX95505-22 MFLX95505-44 MFLX95505-42 MFLX95505-20 MFLX95505-08 MFLX95505-28 MFLX95505-06 MFLX95505-18 MFLX95505-04 MFLX95505-26 MFLX95505-16
Masterflex® Transfer Tubing, DEHP-Free Flexible PVC, Avantor®
Tubing Transfer Tubing
As flexible as you need it.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
Applications: Analytical instruments, general laboratory. Use for transfer of liquids, gases, and inorganic chemicals found in lab, paint, pigment, and soap dispensing applications.
Characteristics:Nontoxic DEHP-free formulation. Flexible, small bend radii. Transparent; allows for visual inspection and flow monitoring.
Certifications: FDA compliant, Proposition 65, REACH
Temperature range: -58 to 165 °F (-50 to 73 °C)
Sterilize: Autoclave.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty