Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, Solva™, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
SC0300 SC0286 SC0290 SC0289 SCO296 3 STOP SC0301 SC0302 SC0288 SC0293 SC0299 SC0294 95605-103-STOP SOLVA SC0298 SC0295 SC0296 SC0291 SC0287 SC0297 SC0292
MFLX95605-18EA 67 USD InStock
MFLX95605-18 MFLX95605-28 MFLX95605-38 MFLX95605-44 MFLX95605-22 MFLX95605-32 MFLX95605-10 MFLX95605-42 MFLX95605-30 MFLX95604-36 MFLX95605-48 MFLX95605-26 MFLX95605-36 MFLX95605-14 MFLX95605-24 MFLX95605-46 MFLX95605-12 MFLX95605-34 MFLX95605-40
Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, Solva™, Avantor®
Tubing Peristaltic Pump Tubing
Designed for optimal operation in your Ismatec pumps.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
To complete your Ismatec® pumping system, order three-stop color-coded tubing and extension tubing. Tubing comes in various formulations for almost any pumping application.
Three pre-attached tubing stops give you two sections—when one section of tubing fatigues, a fresh section is ready to use. The stops on the three-stop tubing are color-coded according to tubing ID. Use three-stop tubing with any of the Ismatec® minicartridge pumps and pump heads. Minicartridges withstand pressures up to 22 psi.
The PVC Solvent/Hydrocarbon tubing is resistant to ozone and UV light, resistant to cracking, swelling, and hardening. It is ideal for chromatography applications and for the transfer of fuels, distillates, lubricants, and glycol-based coolants. It features a yellow, translucent appearance, has a temperature range of –30 to 85 °C, and has a pumping life of approximately 1000 hours at 100 rpm.
The overall length of PVC Solvent/Hydrocarbon 95605-series stop tubing is 16" (406 mm). The legacy Ismatec® stop tubing was 400 mm long.
Order extension tubing under "Accessories". For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty