Would you like to log in again?
Masterflex® Plastic Tubing Clamp Ties, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
CTT60R0HSC3 CTT60R9C2
MFLX95615-10EA 49.1 USD InStock
MFLX95615-10 MFLX95615-00
Masterflex® Plastic Tubing Clamp Ties, Avantor®
Ties
Special curved clamp tie head ensures tight seal.
  • Ideal for general-purpose, single-use applications
Perfect for flexible fuel line, water line, and vacuum line applications. Ties measure 8" L x 0.18" W; maximum diameter of 1-3/4".
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty