Would you like to log in again?
Chemfluor® 367 Transfer Pump Tubing
Supplier: ChemFluor
TSC367-0375-062-50 TSC367-0125-031-50 TSC367-0250-062-50 TSC367-0312-031-50 TSC367-0500-062-50 TSC367-0250-047-50
MFLX95638-04EA 364 USD InStock
MFLX95638-04 MFLX95638-02 MFLX95638-01 MFLX95638-00 MFLX95638-06 MFLX95638-05
Chemfluor® 367 Transfer Pump Tubing
Tubing Transfer Tubing
Meets the demands of various industries.
  • Ideal for general-purpose, single-use, and extended fluid path applications
Applications: Pharmaceutical, semiconductor, electrical, and environmental applications. Excellent for processes dependent on maintaining fluid integrity and high purity.
Characteristics: Exceptionally smooth surface, very low extractables with deionized water and acids, excellent chemical resistance and mechanical properties, low counts of trace metals, superior clarity.
Certification: FDA compliant.
Temperature range: -400 to 450 °F (-240 to 232 °C)
Sterilize: With ethylene oxide, autoclave, or dry heat.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty