Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tygon® 3350 Platinum-Cured Silicone Tubing

Supplier: VWR International

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

F00014 F00003 F00004 F00005 F00007 F00011 F00021 F00020 F00002 F00010 F00006 F00026 F00028 F00015 F00029 F00017 F00009 F00001 F00019 F00018
MFLX95702-09EA 384 USD InStock
MFLX95702-09 MFLX95702-18 MFLX95702-19 MFLX95702-08 MFLX95702-05 MFLX95702-27 MFLX95702-16 MFLX95702-06 MFLX95702-28 MFLX95702-17 MFLX95702-03 MFLX95702-25 MFLX95702-14 MFLX95702-04 MFLX95702-01 MFLX95702-13 MFLX95702-02 MFLX95702-10 MFLX95702-00 MFLX95702-20
Tygon® 3350 Platinum-Cured Silicone Tubing
Tubing Transfer Tubing
Reduced particle entrapment with ultra-smooth inner surface.

  • Minimal extractables help maintain fluid integrity

Formulation: TYGON® 3350 silicone, platinum cured.
Applications: Pharmaceutical, cosmetic, industrial, and auto-analysis applications.
Characteristics: Ultra-smooth inner surface reduces particle entrapment and minimizes bacterial growth. Good for mild to medium concentration bases, salts, and alcohols; is odorless, tasteless, and non-toxic. Transparent.
Certification: Elastomer is FDA-compliant (21 CFR 177.2600); meets USP Class VI requirements, NSF criteria and EP 3.2.9 requirements. Tubing is manufactured according to GMP.
Temperature range: –112 to 392 °F (–80 to 200 °C)
Sterilize: With ethylene oxide, gamma irradiation, or autoclave for 30 min, 15 psi.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty