Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Masterflex® Transfer Tubing, Tygon® SE-200, Non-DEHP FEP-Lined, Avantor®

Supplier: Avantor Fluid Handling
Masterflex®
AJD00007-CP AJD00053-CP AJD00017-CP AJD00038-CP AJD00012-CP AJD00028-CP AJD04007-CP AJD00002-CP
MFLX95713-60EA 936 USD InStock
MFLX95713-60 MFLX95713-00 MFLX95713-10 MFLX95713-01 MFLX95713-50 MFLX95713-20 MFLX95713-41 MFLX95713-30
Masterflex® Transfer Tubing, Tygon® SE-200, Non-DEHP FEP-Lined, Avantor®
Tubing Transfer Tubing
Keep samples free of odors and taste.

  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application

Formulation: Tygon® SE-200
Applications: Chemical and pharmaceutical processing, beverage dispensing, paint and solvent production, semiconductor processing
Characteristics: Bio-based, phthalate-free. Flexible, chemically resistant, inert, non-contaminating. Odorless, nontoxic, and taste free. Clear
Certifications: FDA compliant for contact food products
Temperature range: -49 to 165 °F (-45 to 74 °C)
Sterilize: Ethylene oxide

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty