Would you like to log in again?
Masterflex® L/S® Sanitary Fitting Pump Tubing Assembly, PharMed® BPT, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
96103-25 96102-17 96103-18 96102-16 96102-36 96103-36 96103-15 96103-16 96102-18 96102-24 96103-14 96103-24 96102-25 96103-17 96102-14 96102-35 96102-13 96103-13 96102-15
MFLX96103-24EA 229 USD InStock
MFLX96103-24 MFLX96103-13 MFLX96103-25 MFLX96103-36 MFLX96102-24 MFLX96103-14 MFLX96102-35 MFLX96102-13 MFLX96102-36 MFLX96103-15 MFLX96102-25 MFLX96102-14 MFLX96103-16 MFLX96102-15 MFLX96102-16 MFLX96103-17 MFLX96102-17 MFLX96103-18 MFLX96102-18
Masterflex® L/S® Sanitary Fitting Pump Tubing Assembly, PharMed® BPT, Avantor®
Tubing Peristaltic Pump Tubing
1/2" mini-clamp sanitary fittings are molded onto each end of the assembly for easy implementation into your high-purity system.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
Applications: Cell and tissue culture work, pharmaceutical, light-sensitive samples
Characteristics: Nontoxic, nonhemolytic, extremely low permeability, resists acids, bases, and oxidizing agents; heat-sealable, bondable, formable, opaque beige
Requires sanitary gaskets for both connections to ensure quality seal
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty