Would you like to log in again?
Masterflex® Ismatec® Pump Tubing, GORE® STA-PURE® Series PFL Single Elements, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
AA14FE / ISMATEC EC24FE / ISMATEC XZ14FE / ISMATEC CA12FE / ISMATEC DB24FE / ISMATEC YB14FE / ISMATEC ZA14FE / ISMATEC ZA12FE / ISMATEC CB14FE / ISMATEC AB14FE / ISMATEC BA14FE / ISMATEC DC24FE / ISMATEC BA12FE / ISMATEC AA12FE / ISMATEC BB14FE / ISMATEC YA14FE / ISMATEC YA12FE / ISMATEC ZB14FE / ISMATEC
MFLX96252-10EA 949 USD InStock
MFLX96252-10 MFLX96252-30 MFLX96252-40 MFLX96252-14 MFLX96252-02 MFLX96252-24 MFLX96252-12 MFLX96252-34 MFLX96252-00 MFLX96252-22 MFLX96252-18 MFLX96252-06 MFLX96252-28 MFLX96252-16 MFLX96252-38 MFLX96252-26 MFLX96252-04 MFLX96252-08
Masterflex® Ismatec® Pump Tubing, GORE® STA-PURE® Series PFL Single Elements, Avantor®
Tubing Peristaltic Pump Tubing
Excellent choice for high pressure, low spallation peristaltic tubing applications.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Engineered for long life in peristaltic pump applications
  • Unique composite of expanded PTFE and platinum-cured perfluoroelastomer
Applications: Chemical, critical pharmaceutical, and biopharma processing applications
Characteristics: High resilience and strength, low spallation and gas permeability. Opaque.
Certifications: USP Class VI compliant, REACH and RoHS compliant, and manufactured in Class 7 (ISO 146441-1) cleanroom, under cGMP certified to ISO 13485 and 15378.
Temperature range: -20 to 80 °C
Sterilize: Autoclave, CIP/SIP
The Ismatec® GORE® STA-PURE® Series PFL pump tubing is designed to withstand pumping aggressive chemicals with a composite structure made of platinum-cured perfluoroelastomer and ePTFE. Long-term testing showed pumping common organic solvents (acetic acid, acetone, and ethanol) at 10 psig did not reduce the pumping life of the tubing compared with pumping deionized water at the same pressure. Single element and double-Y element versions available.
An engineering questionnaire must be completed before the initial order can be filled.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty