Would you like to log in again?
Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, Chem-Durance® Bio, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
A055202-EP-CP A055218-EP-CP A055210-EP-CP A055205-EP-CP A055219-EP-CP A055212-EP-CP A055215-EP-CP A055208-EP-CP 06328-36 FROM 96328-36 A055207-EP-CP
MFLX96328-30EA 189.55 USD InStock
MFLX96328-30 MFLX96328-42 MFLX06328-36 MFLX96328-34 MFLX96328-24 MFLX96328-14 MFLX96328-36 MFLX96328-48 MFLX96328-26 MFLX96328-18
Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, Chem-Durance® Bio, Avantor®
Tubing Peristaltic Pump Tubing
Designed for optimal operation in your Ismatec pumps.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Long service life - Outstanding fatigue resistance due to flexing!
  • Suitable for applications using acetone, methyl-ethyl-ketone (MEK), and other highly aggressive chemicals
To complete your Ismatec® pumping system, order three-stop color-coded tubing and extension tubing. Tubing comes in various formulations for almost any pumping application.
Three pre-attached tubing stops give you two sections—when one section of tubing fatigues, a fresh section is ready to use. The stops on the three-stop tubing are color-coded according to tubing ID. Use three-stop tubing with any of the Ismatec® minicartridge pumps and pump heads. Minicartridges withstand pressures up to 22 psi.
Chem-Durance® Bio tubing is opaque in color and features a temperature range of -70 to 52 °C. Tubing life at 100 rpm is approximately 1000 hours.
The overall length of Chem-Durance® Bio 96327 and 96328-series stop tubing is 16" (406 mm). The legacy Ismatec® stop tubing was 381 mm long.
Order extension tubing under "Accessories". For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty