Would you like to log in again?
Masterflex® I/P® Spooled Pump Tubing, Platinum-Cured Silicone, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
3071651 3071669 3071618
MFLX96403-73CS 0 USD InStock
MFLX96403-73 MFLX96403-82 MFLX96403-26
Masterflex® I/P® Spooled Pump Tubing, Platinum-Cured Silicone, Avantor®
Tubing Peristaltic Pump Tubing
Reduce the risk of particulate entrapment with smooth inner surface.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory Approvals to match your unique application
  • No leachable additives protects sensitive and costly fluids
Applications: Pharmaceutical, biotech, peristaltic pumping, and food applications
Characteristics: Excellent heat resistance, low compression set, odorless, nontoxic, translucent
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty