Would you like to log in again?
Masterflex® I/P® Precision Pump Tubing, FDA-Compliant Viton®, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
96412-73 96412-26
MFLX96412-73CS 829 USD InStock
MFLX96412-73 MFLX96412-26
Masterflex® I/P® Precision Pump Tubing, FDA-Compliant Viton®, Avantor®
Tubing Peristaltic Pump Tubing
Safely pump harsh chemicals.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory Approvals to match your unique application
Masterflex® FDA-Compliant Viton® Precision pump tubing is designed for enhanced chemical compatibility at elevated temperatures. Tubing tolerates the transfer of the harshest chemicals such as corrosive solvents and aliphatic and aromatic hydrocarbons. The ultra-low gas permeability maintains integrity of fluid while protecting the user and environment.
Ideal for where FDA compliance and superior chemical compatibility are required.
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty