Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Masterflex® Ismatec® Microbore Pump Tubing, Puri-Flex®, Avantor®

Supplier: Avantor Fluid Handling
Masterflex®
96418-22(1PK) 96418-48 96418-10(1PK) 96418-34(1PK) 96418-24(1PK) 96418-40(1PK) 96418-46(1PK) 96418-12(1PK) 96418-26(1PK) 96418-14 96418-30(1PK) 96418-16(1PK) 96418-44(1PK) 96418-28(1PK) 96418-32(1PK) 96418-18(1PK) 96418-42(1PK) 96418-36(1PK)
MFLX96418-14EA 85 USD InStock
MFLX96418-14 MFLX96418-36 MFLX96418-24 MFLX96418-46 MFLX96418-34 MFLX96418-12 MFLX96418-22 MFLX96418-44 MFLX96418-10 MFLX96418-32 MFLX96418-42 MFLX96418-30 MFLX96418-40 MFLX96418-18 MFLX96418-28 MFLX96418-16 MFLX96418-48 MFLX96418-26
Masterflex® Ismatec® Microbore Pump Tubing, Puri-Flex®, Avantor®
Tubing Peristaltic Pump Tubing
Long life tubing for pharma and bioprocess applications.

  • Tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Regulatory approvals to match your unique application

Applications: General-purpose lab use requiring high performance and long life
Characteristics: Exceptional tensile and tear strength, good biocompatibility, low gas permeability, smooth surface, translucent
Certifications: FDA-compliant (21 CFR 177.2600), meets USP Class VI requirements, manufactured according to GMP
Temperature range: –58 to 275 °F (–50 to 135 °C)
Sterilize: Autoclave, ethylene oxide, or gamma irradiation

For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty