Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Versilon™ 2001 Transfer Tubing

Supplier: Saint Gobain Life Sciences
AE300053 AE300027 AE300017 AE300012 AE300064 AE300038 AE300007 AE300046 AE300003 AE300022
MFLX96430-02EA 214 USD InStock
MFLX96430-02 MFLX96430-14 MFLX96430-04 MFLX96430-16 MFLX96430-06 MFLX96430-18 MFLX96430-08 MFLX96430-10 MFLX96430-00 MFLX96430-12
Versilon™ 2001 Transfer Tubing
Tubing Transfer Tubing
Ideal for chemical transfer applications.

  • Global market leader in range of formulations for most extensive chemical compatibility

Formulation: Versilon™ 2001
Applications: Soap and detergent dispensing, water purification lines, food contact applications and chemical transfer.
Characteristics: Plasticizer-free, translucent, excellent chemical compatibility to weak and medium acids and bases, salts and alcohols.
Certification:FDA compliant for food contact
Temperature range:-108F to 135F (-78 to 57C)
Sterilize: With ethylene oxide, gamma irradiation or steam at 30 minutes at 15 psi (1.0 bar).

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty