Would you like to log in again?
Masterflex® Ismatec® Pump Tubing, 2-Stop Microbore, Tygon® S3™ E-LFL, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
A055518-EP-CP-1.60 A055516-EP-CP A055507-EP-CP A055511-EP-CP A055501-EP-CP A055522-EP-CP-2.20 A055515-EP-CP-1.37 A055521-EP-CP-2.62 A055505-EP-CP A055520-EP-CP A055521-EP-CP A055503-EP-CP A055514-EP-CP A055513-EP-CP A055517-EP-CP A055524-EP-CP A055509-EP-CP A055508-EP-CP A055502-EP-CP
MFLX96449-40EA 152 USD InStock
MFLX96449-40 MFLX96449-30 MFLX96449-22 MFLX96449-33 MFLX96449-34 MFLX96449-12 MFLX96449-42 MFLX96449-31 MFLX96449-10 MFLX96449-43 MFLX96449-37 MFLX96449-48 MFLX96449-26 MFLX96449-24 MFLX96449-47 MFLX96449-14 MFLX96449-36 MFLX96449-28 MFLX96449-18
Masterflex® Ismatec® Pump Tubing, 2-Stop Microbore, Tygon® S3™ E-LFL, Avantor®
Tubing Peristaltic Pump Tubing
Designed for optimal operation in your Ismatec pumps.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Longest tubing life of all Tygon tubing formulations
  • Broad chemical compatibility
  • Clear for easy flow monitoring
To complete your Ismatec® pumping system, order two-stop color-coded and extension tubing. Tubing comes in various formulations for almost any pumping application. The stops on the two-stop tubing are color-coded according to tubing ID. Use this tubing with any of the Ismatec® standard cartridge pumps and pump heads. Cartridges withstand pressures up to 45 psi (2.9 bar).
The overall length of E-LFL 96449-series stop tubing is 16" (406 mm). The legacy Ismatec® stop tubing was 400 mm long.
Order extension tubing under "Accessories". For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty