Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Masterflex® Ismatec® Pump Tubing, 2-Stop Microbore, Tygon® S3™ E-Food, Avantor®

Supplier: Avantor Fluid Handling

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Masterflex®
A056513-EP-CP A056516-EP-CP A056521-EP-CP A056508-EP-CP A056505-EP-CP A056524-EP-CP A056502-EP-CP
MFLX96457-26EA 103 USD InStock
MFLX96457-26 MFLX96457-48 MFLX96457-34 MFLX96457-12 MFLX96457-42 MFLX96457-30 MFLX96457-18
Masterflex® Ismatec® Pump Tubing, 2-Stop Microbore, Tygon® S3™ E-Food, Avantor®
Tubing Peristaltic Pump Tubing
Long service life - Outstanding fatigue resistance due to flexing!

  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Specially designed for use with food applications
  • Extremely smooth bore

Appearance: Opaque
Temperature: -70 to 52 °C
Life at 100 rpm: 1000 hrs

The first of a series of two-stop extension tubing options is below under Accessories. See all of our two-stop extension tubing formulations or contact our Application Specialists for personal assistance.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty