Would you like to log in again?
Masterflex® Ismatec® Pump Tubing, 2-Stop Microbore, Tygon® S3™ E-Lab, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
A053405-EP-CP A053408-EP-CP A053406-EP-CP A053401-EP-CP A053402-EP-CP A053410-EP-CP A053404-EP-CP A053411-EP-CP A053422-EP-CP A053407-EP-CP A053418-EP-CP A053415-EP-CP A053403-EP-CP A053421-EP-CP A053400-EP-CP A053416-EP-CP A053419-EP-CP A053425-EP-CP A053420-EP-CP A053412-EP-CP A053424-EP-CP A053409-EP-CP A053413-EP-CP A053414-EP-CP A053423-EP-CP A053417-EP-CP
MFLX96460-09EA 128 USD InStock
MFLX96460-09 MFLX96460-29 MFLX96460-24 MFLX96460-46 MFLX96460-44 MFLX96460-22 MFLX96460-27 MFLX96460-49 MFLX96460-28 MFLX96460-48 MFLX96460-26 MFLX96460-30 MFLX96460-31 MFLX96460-18 MFLX96460-19 MFLX96460-12 MFLX96460-34 MFLX96460-10 MFLX96460-32 MFLX96460-16 MFLX96460-38 MFLX96460-39 MFLX96460-14 MFLX96460-36 MFLX96460-42 MFLX96460-40
Masterflex® Ismatec® Pump Tubing, 2-Stop Microbore, Tygon® S3™ E-Lab, Avantor®
Tubing Peristaltic Pump Tubing
Designed for optimal operation in your Ismatec pumps.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Ideal for general laboratory applications
  • Clear for easy sample viewing
Appearance: Opaque
Temperature: -70 to 52 °C
Life at 100 rpm: 1000 hrs
The overall length of E-Lab (E-3603) 96460-series stop tubing is 16" (406 mm). The legacy Ismatec® stop tubing was 400 mm long.
Order extension tubing under "Accessories". For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty