Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Masterflex® Transfer Tubing, Cleargreen® Phthalate and DEHP-Free PVC, Avantor®

Supplier: Avantor Fluid Handling

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Masterflex®
CG-500X625-50 CG-375X625-50 CG-312X437-50 CG-125X250-50 CG-1000X1250-50 CG-250X500-50 CG-375X500-50 CG-500X750-50 CG-750X1000-50 CG-625X875-50 CG-250X375-50 CG-187X312-50
MFLX96605-41EA 194 USD InStock
MFLX96605-41 MFLX96605-43 MFLX96605-20 MFLX96605-31 MFLX96605-33 MFLX96605-22 MFLX96605-35 MFLX96605-24 MFLX96605-27 MFLX96605-37 MFLX96605-29 MFLX96605-39
Masterflex® Transfer Tubing, Cleargreen® Phthalate and DEHP-Free PVC, Avantor®
Tubing Transfer Tubing
Eliminate odor transferring to sample.

  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application

Characteristics: Translucent, odorless, low water absorption, low extractables
Certified compliant FDA compliant, USP Class VI, 3A compliant, Reach compliant and RoHS compliant.
Temperature range: -58 to 165 °F (-50 to 74 °C)
Sterilize: Autoclave or ethylene oxide.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty