Would you like to log in again?
Masterflex® Transfer Tubing, PharMed® BPT, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
AY242046-CP AY242005 AY242007-CP AY242019-CP AY242027 AY242012-CP AY242605-CP AY242022-CP AY242006 AY242002 AY242606-CP AY242038-CP AY242053-CP AY242003-CP AY242029-CP AY242017-CP
MFLX96880-20EA 540 USD InStock
MFLX96880-20 MFLX96880-30 MFLX96880-00 MFLX96880-22 MFLX96880-10 MFLX96880-02 MFLX96880-12 MFLX96880-15 MFLX96880-04 MFLX96880-14 MFLX96880-03 MFLX96880-06 MFLX96880-05 MFLX96880-16 MFLX96880-08 MFLX96880-18
Masterflex® Transfer Tubing, PharMed® BPT, Avantor®
Tubing Transfer Tubing
Protect your light-sensitive samples.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
This tubing is ideal for general laboratory use. They offer long life, won't age or oxidize, fits tightly to glass and metal, bends to small radius curves, are nontoxic. Made according to GMP. Temperature range is -51 to 270 °F (-51 to 132 °C). Can be sterilized with ethylene oxide, gamma, or autoclave.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty