Would you like to log in again?
Masterflex® Ismatec® Pump Tubing with Flared Ends, 2-Stop Microbore, Puri-Clear™ LL, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
97616-14 116-0549-03F 97616-22 116-0549-07F 97616-10 97616-26 116-0549-05F 116-0549-09F-CP 97616-30 116-0549-01F
MFLX97616-12EA 74 USD InStock
MFLX97616-12 MFLX97616-24 MFLX97616-10 MFLX97616-22 MFLX97616-30 MFLX97616-09 MFLX97616-18 MFLX97616-28 MFLX97616-14 MFLX97616-26
Masterflex® Ismatec® Pump Tubing with Flared Ends, 2-Stop Microbore, Puri-Clear™ LL, Avantor®
Tubing Peristaltic Pump Tubing
Flared ends for easy extension tubing assembly.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Pthalate-free, USP Class VI PVC
  • Very low Gas Permeability
Two-Stop Flared PVC Color-coded Autoanalysis Tubing for Ismatec® Pumps is USP Class VI tubing that is Phthalate-free. It has a long pumping life and exhibits a wide range of acid and chemical resistance, thus making it an ideal choice for Pharmaceutical industries. Plasticizer is very tightly bonded into the material, preventing any leaching of component chemicals into the medium being transported, maintaining its sterility. The tubing is transparent, which allows visual observation of tubing contents. It also offers very low gas permeability and can withstand high pressure contents.
Tubing is also called TAAT / microbore and auto analysis.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty