Would you like to log in again?
Masterflex® Ismatec® Pump Tubing, 2-Stop Microbore, Tygon® LMT-55, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
ALH053401-EP-CP ALH053413-EP-CP ALH053405-EP-CP ALH053407-EP-CP ALH053417-EP-CP ALH053421-EP-CP ALH053403-EP-CP ALH053425-EP-CP ALH053411-EP-CP ALH053415-EP-CP ALH053416-EP-CP ALH053420-EP-CP ALH053424-EP-CP ALH053419-EP-CP ALH053423-EP-CP 070534-10I-N ALH053418-EP-CP ALH053422-EP-CP ALH053412-EP-CP ALH053409-EP-CP ALH053408-EP-CP ALH053404-EP-CP ALH053414-EP-CP ALH053400-EP-CP
MFLX97618-48EA 142 USD InStock
MFLX97618-48 MFLX97618-26 MFLX97618-36 MFLX97618-14 MFLX97618-46 MFLX97618-24 MFLX97618-34 MFLX97618-22 MFLX97618-44 MFLX97618-10 MFLX97618-32 MFLX97618-42 MFLX97618-31 MFLX97618-30 MFLX97618-09 MFLX97618-18 MFLX97618-29 MFLX97618-28 MFLX97618-39 MFLX97618-49 MFLX97618-27 MFLX97618-38 MFLX97618-16 MFLX97618-40
Masterflex® Ismatec® Pump Tubing, 2-Stop Microbore, Tygon® LMT-55, Avantor®
Tubing Peristaltic Pump Tubing
Designed for optimal operation in your Ismatec pumps.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
The overall length of LMT-55 97618-series stop tubing is 16" (406 mm). The legacy Ismatec® stop tubing was 400 mm long.
Order extension tubing under "Accessories". For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty