Would you like to log in again?
Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, Tygon® LMT-55, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
070535-05I-N 070535-11I-N 070535-22I-N 070535-17I-N 070535-02I-N 070535-01I-N 070535-20I-N 070535-08I-N 070535-24I-N 070535-03I-N 070535-14I-N 070535-06I-N 070535-15I-N 070535-10I-N 070535-07I-N 070535-16I-N 070535-25I-N 070535-00I-N 070535-23I-N 070535-12I-N 070535-04I-N 070535-13I-N 070535-21I-N 070535-09I-N 070535-19I-N 070535-18I-N
MFLX97619-27EA 144 USD InStock
MFLX97619-27 MFLX97619-49 MFLX97619-48 MFLX97619-26 MFLX97619-24 MFLX97619-46 MFLX97619-44 MFLX97619-22 MFLX97619-42 MFLX97619-09 MFLX97619-29 MFLX97619-28 MFLX97619-40 MFLX97619-16 MFLX97619-38 MFLX97619-36 MFLX97619-14 MFLX97619-12 MFLX97619-34 MFLX97619-10 MFLX97619-32 MFLX97619-31 MFLX97619-19 MFLX97619-18 MFLX97619-39 MFLX97619-30
Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, Tygon® LMT-55, Avantor®
Tubing Peristaltic Pump Tubing
Designed for optimal operation in your Ismatec pumps.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
To complete your Ismatec® pumping system, order three-stop color-coded tubing and extension tubing. Tubing comes in various formulations for almost any pumping application.
Three pre-attached tubing stops give you two sections—when one section of tubing fatigues, a fresh section is ready to use. The stops on the three-stop tubing are color-coded according to tubing ID. Use three-stop tubing with any of the Ismatec® minicartridge pumps and pump heads. Minicartridges withstand pressures up to 22 psi.
The Tygon® LMT-55 features a clear appearance, has a temperature range of -58 °F to 165 °F, and a smooth, polished inner wall prevents buildup.
The overall length of LMT-55 97619-series stop tubing is 16" (406 mm). The legacy Ismatec® stop tubing was 400 mm long.
Order extension tubing under "Accessories". For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty