Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Masterflex® Ismatec® Pump Tubing, 2-Stop Microbore, FDA-Compliant Viton®, Avantor®

Supplier: Avantor Fluid Handling

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Masterflex®
T97628-18 97628-42 97628-26 T97628-40 96428-48 96428-26 T97628-32 T97628-28 T97628-44 96428-42 97628-46 97628-48 T97628-36 97628-30 97628-38 T97628-24 97628-34 T97628-22 96428-34
MFLX96428-34EA 330 USD InStock
MFLX96428-34 MFLX96428-42 MFLX96428-48 MFLX96428-26 MFLX97628-34 MFLX97628-44 MFLX97628-22 MFLX97628-36 MFLX97628-46 MFLX97628-24 MFLX97628-30 MFLX97628-40 MFLX97628-32 MFLX97628-42 MFLX97628-38 MFLX97628-26 MFLX97628-48 MFLX97628-18 MFLX97628-28
Masterflex® Ismatec® Pump Tubing, 2-Stop Microbore, FDA-Compliant Viton®, Avantor®
Tubing Peristaltic Pump Tubing
Designed for optimal operation in your Ismatec pumps.

  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application

Applications: Acid and solvent transfer; high-temperature work
Characteristics: Opaque black
Resin certification: Tubing is manufactured according to GMP
Temperature range: -8 to 400 °F (-13 to 204 °C)
Sterilize: At 480 °F (249 °C) for 16 hours in a circulating hot air oven
The overall length of Viton® 97628-series stop tubing is 12" (304 mm). The legacy Ismatec® stop tubing was 180 mm long.
Order extension tubing under "Accessories". For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty