Would you like to log in again?
Evsco Valves Manual and Actuated Accessories
Supplier: Evsco
UMB-1 UMB-2
MFLX98150-50EA 23.2 USD InStock
MFLX98150-50 MFLX98150-51
Evsco Valves Manual and Actuated Accessories
Valves Elliptical Valves
Safely mount your plastic elliptical valves.
  • Optimize performance of your Evsco Valves
Constucted of durable polypropylene, these mounting brackets are ideal for making your valve more versitile. These mounting brackets allow for wall, panel, or undermount.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty