Would you like to log in again?
Masterflex® Solenoid-Operated Pinch Valves, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
075P2NO-24-01 075P2NC-24-02 100P2NO-12-05 98302-06 0753MP-12-02 075P2NC-12-01 100P2NO-24-05 98302-02 0753MP-24-01 075P2NO-24-02 98302-16 075P2NO-12-02 075P2NC-24-01 075P3MP-24-02S 0753MP-12-01 100P3MP-12-05 075P2NO-12-01 98302-54
MFLX98302-06EA 185 USD InStock
MFLX98302-06 MFLX98302-26 MFLX98302-48 MFLX98302-16 MFLX98302-02 MFLX98302-46 MFLX98302-36 MFLX98302-00 MFLX98302-22 MFLX98302-12 MFLX98302-42 MFLX98302-20 MFLX98302-32 MFLX98302-10 MFLX98302-54 MFLX98302-40 MFLX98302-30 MFLX98302-50
Masterflex® Solenoid-Operated Pinch Valves, Avantor®
Valves Solenoid Valves
No contamination of valve.
  • Control flow in your tubing without direct contact between the fluid and the valve
  • Fast response time
  • Low-power consumption and high-cycle life
  • Excellent for sanitary and pharmaceutical applications
Choose from two-way NC, two-way NO, or three-way pinch valves. These are ideal where contamination is an issue, as the fluid only comes in contact with the tubing, not the valve. Tubing replacement is quick and easy, making these suitable for pharmaceutical or sanitary applications. Mounting clip or mounting flange available separately for increased versatility.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty