Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Guanidinium hydrochloride 99.5-101.0%, white powder, NF-GenAR® for biotechnology, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: Guanidinium chlorideGuanidine hydrochlorideGuanidine chlorideAminoformamidine hydrochlorideGuanadine hydrochlorideAminomethanamidine hydrochlorideGuanidine HCI

Management of Change (MOC) category = R

MACR0507-06 MACR0507-F7 MACR0507-F9 MACR0507-F9 MACR0507-25 MACR0507-88 MACR0507-F7 MACR0507-88 MACR0507-04 MACR0507-04 MACR0507-25 MACR0507-06
MK050706EA 2972.31 USD InStock
MK050706 CAMK050788 CAMK0507F7 MK050704 MK0507F9 MK050725 CAMK0507F9 CAMK050725 MK0507F7 CAMK050704 MK050788 CAMK050706
Guanidinium hydrochloride 99.5-101.0%, white powder, NF-GenAR® for biotechnology, Macron Fine Chemicals™
Guanidinium hydrochloride


Formula: NH₂C(=NH)NH₂·HCl
MW: 95.53 g/mol
Melting Pt: 185 °C
Density: 1.354 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013026
CAS Number: 50-01-1
EINECS: 200-002-3
Merck Index: 13,04578
https://us-legacy.vwr.com/stibo/web/std.lang.all/74/24/2297424.jpg
Order Now
Specification Test Results
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Assay (NH₂C(:NH)NH₂·HCl) 99.5 - 101.0 %
Acidity (as HCl) ≤0.01 %
Loss on Drying at 105°C ≤0.5 %
Residue after Ignition ≤0.050 %
Absorbance of a 6M Aqueous Solution - 230 nm ≤0.200
Absorbance of a 6M Aqueous Solution - 260 nm ≤0.030
Absorbance of a 6M Aqueous Solution - 275 nm ≤0.030
RNase Activity (None Detected) Passes Test
DNase Activity (None Detected) Passes Test
Protease Activity (None Detected) Passes Test
Nitrate (NO₃) ≤0.005 %
Sulfate (SO₄) ≤0.005 %
Arsenic and Antimony (as As) ≤0.500 ppm
Trace Impurities - Copper (Cu) ≤0.200 ppm
Cyanide (CN) ≤0.1 ppm
Trace Impurities - Iron (Fe) ≤2.0 ppm
Trace Impurities - Lead (Pb) ≤0.500 ppm
Particulate Matter Passes Test
MANUFACTURED EXCLUSIVELY FOR AVANTOR PERFORMANCE MATERIALS, LLC., BY
BIOSPECTRA, INC., STROUDSBURG, PA

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty