Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phenol 99.0-100.5% (by anhydrous basis), crystals USP, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Carbolic acidHydroxybenzenePhenylalcohol

Management of Change (MOC) category = R

MACR0605-06 MACR0605-06 MACR0605-04 MACR0605-04
MK060504-UCS 5049.65 USD InStock
CAMK060506 MK060504-U CAMK060504 MK060506
Phenol 99.0-100.5% (by anhydrous basis), crystals USP, Macron Fine Chemicals™
Phenol


Formula: C₆H₅OH
MW: 94.11 g/mol
Boiling Pt: 181.8 °C (1013 hPa)
Melting Pt: 40.8 °C
Density: 1.06 g/cm³ (20 °C)
Flash Pt: 81 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002143
CAS Number: 108-95-2
EINECS: 203-632-7
UN: 1671
ADR: 6.1,II
Order Now
Specification Test Results
GMP Manufactured Product
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Assay (C₆H₅OH) (anhydrous basis) 99.0 - 100.5 %
USP - Appearance of Melted Sample Passes Test
USP - Benzene ≤2.0 ppm
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Clarity of Solution and Reaction Passes Test
USP - Congealing Temperature ≥39 °C
USP - Nonvolatile Residue ≤0.05 %
USP - Water (H₂O)(by Karl Fischer titrn) ≤0.5 %
Appearance (colorless to light pink, crystalline mass)
No Class 2 or 3 solvents are expected to be present. The Class 1
solvent Benzene is used as a raw material in a precursor to phenol. The product is tested for any presence of benzene and the result
reported on the Certificate of Analysis.
Product contains about 20 ppm Citric Acid as a preservative.
Elemental Impurities (USP 232) - Information on elemental impurities for
this product is available on the associated Product Regulatory Data
Sheet and elemental impurity profile report.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty