Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Boric acid ≥99.5%, granular NF, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Boric acidBoron hydroxideBoron trihydroxideBoracic acidOrthoboric acid

Management of Change (MOC) category = R

MACR1394-26 MACR1394-20 MACR1394-20
MK139420EA 1457 USD InStock
CAMK1394-26 CAMK139420 MK139420
Boric acid ≥99.5%, granular NF, Macron Fine Chemicals™
ortho-Boric acid
GMP manufactured.


Formula: B(OH)₃
MW: 61.83 g/mol
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 160 °C
Density: 1.44 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011337
CAS Number: 10043-35-3
EINECS: 233-139-2
REACH: 01-2119486683-25
Merck Index: 13,01326
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/54/16640554.jpg
Order Now
Specification Test Results
GMP Manufactured Product
Meets N.F. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
NF - Assay (H₃BO₃) 99.5 - 100.5 %
NF - Completeness of Solution Passes Test
NF - Identification Passes Test
NF - Loss on Drying ≤0.5 %
NF - Solubility in Alcohol Passes Test
Color of Solution Passes Test
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Not for Internal Use
Meets Reagent Specifications for testing USP/NF monographs
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty