Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Benzyl alcohol 98.0-100.5% NF, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: PhenylmethanolBenzenemethanolPhenylcarbinol

Management of Change (MOC) category = R

MACR1403-12 MACR1403-10 MACR1403-25 MACR1403-10 MACR1403-25 MACR1403-19 MACR1403-12 MACR1403-19
MK140319EA 1262.32 USD InStock
MK140319 CAMK140312 CAMK140310 CAMK1403-25 MK1403-25 MK140312 MK140310 CAMK140319
Benzyl alcohol 98.0-100.5% NF, Macron Fine Chemicals™
Benzyl alcohol
Caution: This product is not intended for use in preparing parenteral dosage forms.


Formula: C₆H₅CH₂OH
MW: 108.14 g/mol
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: –15.3 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00004599
CAS Number: 100-51-6
EINECS: 202-859-9
Merck Index: 13,01126
https://us-legacy.vwr.com/stibo/web/std.lang.all/97/87/2299787.jpg
Order Now
Specification Test Results
GMP Manufactured Product
Meets N.F. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
NF - Assay (C₆H₅CH₂OH) 98.0 - 100.5 %
NF - Acidity Passes Test
NF - Clarity of Solution Passes Test
NF - Color of Solution Passes Test
NF - Identification Passes Test
NF - Residue on Evaporation ≤0.05 %
NF - Peroxide Value ≤5
NF - Refractive Index, n²ºD 1.538 - 1.541
NF - Benzaldehyde and Other Related Substances - Benzaldehyde ≤0.15 %
NF - Benzaldehyde and Other Related Substances - Cyclohexylmethanol ≤0.10 %
NF - Benzaldehyde and Other Related Substances - Total Other Peaks Less ≤0.04 %
Than RT of Benzyl Alcohol ≤0.04 %
NF - Benzaldehyde and Other Related Substances - Total Other Peaks ≤0.3 %
Greater Than RT of Benzyl Alcohol ≤0.3 %
NF - Benzene ≤2 ppm
Solubility Passes Test
The Class 1 residual solvent Benzene is likely to be present below 2 ppm. The actual analysis is reported for each lot. The Class 2 solvent Toluene is likely to be present and is process controlled to <30 ppm. No other Class 1, 2 or 3 residual solvents are likely to be present.
This product is not intended for use in preparing parenteral dosage forms.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty