Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Dimethylamino)benzaldehyde, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: Ehrlich's Reagent4-(N,N-Dimethylamino)benzaldehydep-Dimethylamino-Benzaldehydep-(Dimethylamino)benzaldehyde

Management of Change (MOC) category = N

MACR1836-57 MACR1836-57 MACR1836-55 MACR1836-55
MK183655EA 141.75 USD InStock
MK183655 MK183657 CAMK183655 CAMK183657
4-(Dimethylamino)benzaldehyde, AR® ACS, Macron Fine Chemicals™
4-(Dimethylamino)benzaldehyde


Formula: (CH₃)₂NC₆H₄CHO
MW: 149.19 g/mol
Boiling Pt: 176…177 °C (1013 hPa)
Melting Pt: 72 °C
Density: 1.0254 g/cm³ (99.9 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00003381
CAS Number: 100-10-7
EINECS: 202-819-0
Merck Index: 13,03257
https://us-legacy.vwr.com/stibo/web/std.lang.all/62/54/746254.jpg
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Solubility in Alcohol Passes Test
ACS - Appearance (white to yellowish-white crystals or crystalline powder) Passes Test
ACS - Color of HCl Solution Passes Test
ACS - Melting Point 73 - 75 °C
ACS - Residue after Ignition ≤0.1 %
ACS - Solubility in HCl Passes Test
ACS - Color of Alcohol Solution (APHA) ≤60

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty