Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trifluoroacetic acid ≥99%, OR, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: TFA

Management of Change (MOC) category = N

MACR1942-59 MACR1942-59
MK194259EA 403.78 USD InStock
CAMK194259 MK194259
Trifluoroacetic acid ≥99%, OR, Macron Fine Chemicals™
Trifluoroacetic acid


Formula: C₂HF₃O₂
MW: 114.02 g/mol
Boiling Pt: 71.8 °C (1013 hPa)
Melting Pt: –15 °C
Density: 1.490 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004169
CAS Number: 76-05-1
EINECS: 200-929-3
UN: 2699
ADR: 8,I
Merck Index: 12,09812
https://us-legacy.vwr.com/stibo/web/std.lang.all/83/92/728392.jpg
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay (F₃CCOOH) ≥99 %
Appearance (colorless to light straw fuming liquid)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty